Orders LinkContactUsLinkComponent
Ward's® Essentials Stereomicroscope
Ward's® Essentials Stereomicroscope
Stereomicroscope, Cordless, 2×, 4×, Rechargeable LED
Catalog # 470315-512
Supplier:   VWR
Supplier #:  SE306R-P-LED
Ward's® Essentials Stereomicroscope
Catalog # 470315-512
Supplier:  VWR
Ward's® Essentials Stereomicroscope
Ward's® Essentials Stereomicroscope
Stereomicroscope, Cordless, 2×, 4×, Rechargeable LED
Price
$237.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Versatile and durable, this stereomicroscope is ideal for the high school classroom.
  • 20× and 40× magnifications
  • Interchangeable stage plates: translucent and black/white
  • Adjustable interpupillary distance
  • One year limited warranty
  • Rechargeable LED illumination
Features include 45° inclined viewing head, post mount that adjusts the head up or down to accommodate larger objects on stage and easy-adjust collar for tension control.
Specifications
  • Description:
    Stereomicroscope, Cordless, 2×, 4×
  • Illumination:
    Rechargeable LED