Teasing Needle with Chuck
Teasing Needle and Handle, Straight, Chrome plated, 135 mm (5.25")
Catalog # 470315-252
Supplier: DR INSTRUMENTS INC. SE
: 37
Jump to:
Product Details & Documents
Durable and easy to clean.
- Brass chuck
- Replicable needle
Often found only in dissection kits, this teasing needle wont crack or chip like wooden or plastic handles. The needle fits securely into the handle and is held in place with a threaded ring.
: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Description:Teasing Needle and Handle, Straight
- Length:135 mm (5.25")
- Material:Chrome plated



