VWR® Upright Freezer Drawer Rack for 96-Well Microtube Boxes
Catalog # 76051-838
Supplier: VWR International
Specifications
- Width:5⁷⁄₁₆"
- Capacity:24
- Height:9¹¹⁄₁₆"
- Length:21½"
- Cat. no.:76051-838
Specifications
About this item
Designed for use with 96-well microtube boxes.
- Made of corrosion resistant stainless steel
- Security lock devices are available for each rack
Drawer racks allow easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.