4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) solution fluorescent dye, for microbiology
Catalog # 10791-650
Supplier: Genetex
CAS Number:
Specifications
- Size:10 ml
- Cat. no.:10791-650
Specifications
About this item
:
:350.25 g/mol
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681