Refractive Index Liquids, Series E
Catalog # 30617-960
Supplier: Cargille Laboratories
CAS Number:
Specifications
- Description:Series E nD 1.500-1.640 ±0.0002
- Size:30 ml (1 oz.)
- Cat. no.:30617-960
- Supplier No.:1843Y
Specifications
About this item
Refractive index liquids have a broad range of applications in diversified fields.
- Slightly volatile
- Colorless flurocarbon formulation
Adjustment: ±0.0002
Choose from Range nD 1.500 to 1.640.with liquids at 0.0005 intervals.
Ordering information: Required refractive index has to be specified with order.