Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
MitoView™
MitoView™, green mitochondria dye, for fluorescence microscopy
MitoView™, green mitochondria dye, for fluorescence microscopy
MitoView™, green mitochondria dye, for fluorescence microscopy
Catalog # 89139-464
Supplier:
Biotium
CAS Number:
Expand image
MitoView™, green mitochondria dye, for fluorescence microscopy
Catalog # 89139-464
Supplier:
Biotium
Supplier Number:
70054
CAS Number:
Specifications
Size:
20×50 μg
Cat. no.:
89139-464
Supplier No.:
70054
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation