Starch 1% (w/v) 0.5N in aqueous solution stabilized indicator
Catalog # CA11024-420
Supplier: Spectrum Chemical Mfg. Corp.
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:4 L
- Cat. no.:CA11024-420
- Supplier No.:S-572-55003
Specifications
About this item
:
:162.14 g/mol
Storage Temperature:Ambient
CAS Number:9005-25-8
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026