Orders LinkContactUsLinkComponent
pH Buffer Capsules, Oakton® Instruments
pH Buffer Capsules, Oakton® Instruments
pH Buffer Capsules, Oakton® Instruments
Supplier:  Antyila Scientific
Certificates
pH Buffer Capsules, Oakton® Instruments
pH Buffer Capsules, Oakton® Instruments
3 Options Available Below
Product Details & Documents
Fresh pH calibration solution anytime and anywhere.
  • One capsule to 100 ml of deionized or distilled water
  • Dry and compact—easily take them anywhere
  • Unused capsules have a five-year shelf life
  • Each vial contains 10 capsules
Buffer salts inside capsules require 100 mL of deionized or distilled water to produce a clear pH calibration solution accurate to ±0.02 pH (at 25°C) with traceability to NIST standards. Capsules are easier to store and transport than prepared aqueous buffer solutions. Capsules are color-coded for easy identification.
Once dissolved, store buffer solution in an airtight container at room temperature for up to three weeks.
Product Family Options
Product Information
  • Final volume
    pH accuracy
    Reference temperature
    Size
    pH
    Availability
    Price
  • 76325-406
    pH capsules
    pH capsules
    100 ml
    ±0.02
    25 °C
    10 capsules/vial
    4.00
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $41.55
    $41.55
    New Customer Promotional Price
    Pack of 10
       $41.55
    Supplier Number: 35850-12
  • pH capsules
    100 ml
    ±0.02
    25 °C
    10 capsules/vial
    7.00
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $49.78
    $49.78
    New Customer Promotional Price
    Pack of 10
       $49.78
    Supplier Number: 35850-14
  • 76325-410
    pH capsules
    pH capsules
    100 ml
    ±0.02
    25 °C
    10 capsules/vial
    10.00
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $41.55
    $41.55
    New Customer Promotional Price
    Pack of 10
       $41.55
    Supplier Number: 35850-16