Order Entry
Canada
Orders LinkContactUsLinkComponent
Plier, Large Frame, Excelta

Plier, Large Frame, Excelta

Long Grips, 163 mm (6.5")
Catalog # 75880-658
Supplier:   Excelta
Supplier #:  907-ICA
Plier, Large Frame, Excelta
Catalog # 75880-658
Supplier:  Excelta
Plier, Large Frame, Excelta
Plier, Large Frame, Excelta

Long Grips, 163 mm (6.5")

Price
$854.42
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

These pliers form stress relief for thru-hole components.

  • Large frame
  • Long cushioned grips – ESD safe (1010 ohms/sq)

They feature dual leaf springs for smooth action. Model 907-ICA forms stress relief on multiple leads.

Specifications
  • Description:
    Long Grips
  • Overall length:
    163 mm (6.5")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.