Whirl-Pak® Polyurethane Surface Sampling Bags with PolySponge™, Nasco
Product Details & Documents
Make environmental surface sampling faster and easier with Whirl-Pak polyurethane surface sampling bags with premoistened sponge. The durable high-density polyurethane sponge is ready to use and resists tearing and fraying.
- Each sponge is packed in a sterile, sealed write-on bag
- Write-on label makes identifying samples easy and convenient
- Sponge is hydrated with 10 ml of HiCap™ neutralizing broth
- Terminally gamma processed to guarantee sterility (+1 Sterilization)
- Opening Tyvek® Pull Tabs are perpendicular to the opening of the bag: Keeps your hands away for a safe, aseptic collection technique each time
To use the sponge, tear open the top of the Whirl-Pak® bag, squeeze out any excess HiCap™ Neutralizing Broth, swab the surface, insert the sponge back into the bag, fold the top of the bag over three or more times, and bend in the wire tabs to close. Send to a lab for testing.
Shelf life from sterilisation date found on the sterility documentation: 2 years from sterilised date found on your sterility confirmation due to the buffer. 2 year shelf-life with ambient storage at 2 to 30 °C. The bags are packed in a foil liner bag to prevent buffer evaporation.
: Approved by USDA, FDA, EPA and HACCP. Food safe.
Specifications
- Sterility:Sterile
- Capacity:18 oz.
- Description:Sampling Bags with hydrated PolySponge™
- L×W:9×4"
- Thickness:2.5 mil.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



