Order Entry
Canada
Orders LinkContactUsLinkComponent
(S)-(-)-Nicotine solution, Reagent Grade

(S)-(-)-Nicotine solution, Reagent Grade

100 ml
Catalog # 470191-572
Irritant
Environment
Supplier:   Ward's Science
Supplier #:  NN0195-100
(S)-(-)-Nicotine solution, Reagent Grade
Catalog # 470191-572
Supplier:  Ward's Science
CAS Number:  54-11-5
(S)-(-)-Nicotine solution, Reagent Grade
(S)-(-)-Nicotine solution, Reagent Grade

100 ml

CAS Number:  54-11-5
Price
$34.05
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Cigarette Solution / Cigarette Water Extract

Composition:
Water > 99.00%
Sodium benzoate 0.25%
Nicotine
Tar Not known

Synonyms:  (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine
  • CAS Number:54-11-5
  • Formula:C₁₀H₁₄N₂
  • MDL Number:MFCD00006369
  • Molecular Weight:162.23 g/mol
  •  :Packaged under nitrogen.

Documents

Specifications

Specifications

  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.