Orders LinkContactUsLinkComponent
Biox Pipette Stand
Biox Pipette Stand
Pipette Stand
Catalog # 76319-892
Supplier:   TWD Scientific
Supplier #:  BX01-2306
Biox Pipette Stand
Catalog # 76319-892
Supplier:  TWD Scientific
Biox Pipette Stand
Biox Pipette Stand
Pipette Stand
Price
139,45 $
Log in for availability
Each
Product Details & Documents
The pipette stand holds up to 6 single channel pipettes or 1 multi-channel pipette in a linear position.
  • Holds six pipettes
  • Horizontal storage
The universal pipette stand is sturdy and ideal for benchtop storage.
Specifications
  • Description:
    Pipette Stand