ContactUsLinkComponent
VWR® Upright Freezer Drawer Rack for 0.2 ml Tube 96-Well PCR Boxes
VWR® Upright Freezer Drawer Rack for 0.2 ml Tube 96-Well PCR Boxes
  76052-414
undefined
VWR® Upright Freezer Drawer Rack for 0.2 ml Tube 96-Well PCR Boxes
  76052-414
  • Width:
    5⁷⁄₁₆"
  • Capacity:
    48
  • Height:
    12½"
  • Length:
    21¹⁵⁄₁₆"
  • Cat. no.:
    76052-414
Designed for use with 0.2 ml tube 96-well PCR boxes.
  • Made of corrosion resistant stainless steel
  • Each compartment holds two boxes
Drawer racks allow easy access to and storage of samples without having to remove the whole rack from the freezer.