Peroxidase
Catalog # CA80602-318
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- CAS No.:9003-99-0
- Species:Horseradish
- Size:5 KU
- Storage conditions:Store at –20 °C
- Form:Lyophilized solid
- Enzyme name:Peroxidase
- Enzyme activity:≥200 units/mg
- Cat. no.:CA80602-318
- Supplier No.:516531-5KU
Specifications
About this item
Suitable for conjugation to proteins and antibodies and as a cytochemical marker in immunohistochemical staining.