About this item
Proteomix ion-exchange phases have been innovatively developed for achieving high ion-exchange capacity, a breakthrough technology for non-porous resins.
- Uniform polymer beads as the packing support
- Unprecedented separation efficiency, selectivity and resolving power
- Complete selection for analytical, semi-preparative and preparative separations
- Wide particle size selection
- High stability and recovery
- High lot-to-lot reproducibility
- Well suited for UPLC system
These columns have a proprietary surface chemistry specially designed for elimination of non-specific bindings and tunable separation capacity with combination of the pore structure and the controllable surface chemistry. The Proteomix SCX columns are compatible with nonionic and zwitterionic detergents and incompatible with cationic detergents.
Proteomix packing support is composed of a rigid, spherical, highly cross-linked poly (styrene divinylbenzene) (PS/DVB) non-porous bead. The PS/DVB resin surface is grafted with a highly hydrophilic, neutral polymer thin layer with the thickness in the range of nanometer. The hydrophobic resin surface is totally covered by a hydrophilic coating that eliminates non-specific bindings with biological analytes, leading to high efficiency and high recovery separations for biological molecules. On the top of the hydrophilic layer, ion-exchange functional groups are attached via a proprietary chemistry, resulting in a high-capacity ion-exchange layer.
Non-porous Proteomix ion-exchange phases are unique because they combine increased capacity and resolution with their intrinsic advantages of high efficiency and high separation speed. These are especially suitable for high resolution, high efficiency and high recovery separations of proteins, oligonucleotides, peptides, polysaccharides, cell lysates, nanoparticles and nanotubes.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 103058-762
- 103059-174
- 103059-126
- 103059-168
- 103059-166
- 103059-122
- 103059-076
- 103059-164
- 103059-074
- 103059-196
- 103059-184
- 103059-128
- 103059-084
- 103059-082
- 103059-160
- 103059-158
- 103059-282
- 103059-280
- 103059-226
- 103059-162
- 103059-068
- 103059-064
- 103059-062
- Binding capacity:
- ~38 mg/ml
- ~20 mg/ml
- ~60 mg/ml
- ~20 mg/ml
- ~20 mg/ml
- ~38 mg/ml
- ~20 mg/ml
- ~20 mg/ml
- ~20 mg/ml
- ~20 mg/ml
- ~20 mg/ml
- ~60 mg/ml
- ~54 mg/ml
- ~54 mg/ml
- ~20 mg/ml
- ~20 mg/ml
- ~38 mg/ml
- ~38 mg/ml
- ~20 mg/ml
- ~20 mg/ml
- ~60 mg/ml
- ~54 mg/ml
- ~54 mg/ml
- Max. pressure:
- 6000 psi
- 4000 psi
- 12000 psi
- 4000 psi
- 4000 psi
- 6000 psi
- 4000 psi
- 4000 psi
- 4000 psi
- 4000 psi
- 4000 psi
- 12000 psi
- 8000 psi
- 8000 psi
- 4000 psi
- 4000 psi
- 6000 psi
- 6000 psi
- 4000 psi
- 4000 psi
- 12000 psi
- 8000 psi
- 8000 psi
- pH range:
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C