Bulk-Pak™ Serological Pipettes, Bellco Glass
Supplier: Bellco Glass
Certificates
About this item
Type 1 Class B Borosilicate Glass.
- Calibrated tips
Plain top promotes rapid even flow with a calibrated tip. All Bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings.
Customers Who Bought This Also Bought