Sodium thiosulfate concentrate for dilution to 1L, 0.1 M Na2S2O3 (0.1N) volumetric solution, Fluka™
Catalog # BJ38200-1EA
Supplier: Honeywell Research Chemicals
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Ampoule
- Size:1 Each
- Cat. no.:BJ38200-1EA
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:158.11 g/mol
Boiling Pt:100 °C
Flash Pt:Does not flash
Flash Pt: Does not flash
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499