Sodium chloride concentrate for dilution to 1L, 0.1 M NaCl (0.1N) volumetric solution, Fluka™
Catalog # BJ32038-1EA
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Size:1 Each
- Cat. no.:BJ32038-1EA
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:58.44 g/mol
Density:1.00 g/cm³
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477