
Potassium thiocyanate concentrate for dilution to 1L, 0.1 M KSCN volumetric solution, Fluka™
Catalog # BJ38140-1EA
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Size:1 Each
- Cat. no.:BJ38140-1EA
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
Density:1 g/cm³ (25 °C)
:97.18 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413