Specifications
- Description:Telescopic Handle, Maxi, Color Coded
- Material:Aluminum
- Size:9'–15'
- Cat. no.:76318-350
Specifications
About this item
Aluminum extendable handle extends to 9'–15' to make reaching those hard to reach areas easy. Lightweight, ergonomic and durable, this handle comes with color coding tabs to reduce cross-contamination. Can be used with all the CE Above The Floor Tools™.
- Designed for use with all CE Above The Floor Tools™
- Aluminum construction
- Autoclavable
- Color coded rings (blue, red, yellow, green) to reduce cross-contamination
Customized tools for special surfaces. The easiest, most effective way to clean pipes, soft walls curtains, tanks and other hard to reach areas in Controlled Environments.
A new line of specialty above-the-floor tools by Vileda Professional®. This line is ideal for all surfaces that are hard or difficult to reach, clean, and disinfect. Specialty tools use Controlled Environment compatible disposable microfiber sleeves that are already validated materials and are an extension of a current CE floor cleaning system.