Micro Fluorescence Cuvettes with PTFE Cover, FireflySci
Supplier: FIREFLYSCI, INC.
Certificates
About this item
High-quality micro cuvettes in a variety of material types and are a great value.
- Fluorescence cells with micro capacities (0.350 to 0.700 ml)
- Comes in several different materials including optical glass, UV quartz and IR quartz
- Lightpaths of 5 mm available with heights of 33 and 45 mm
- Four windows and base optically polished
- Includes PTFE cover
Firefly's micro fluorometer cuvette with PTFE cover. Made by hand, ensuring high quality optical polishing with no defects. Each cell goes through a vigorous inspection process before leaving the factory. Fire-fused to ensure that cuvettes are robust and durable.
Customers Who Bought This Also Bought