ContactUsLinkComponent
HgX® Mercury Decontaminant Powder, Action Technologies
Catalog # 80701-498
Supplier:  ACTON TECHNOLOGIES
CAS Number:  
HgX® Mercury Decontaminant Powder, Action Technologies
Catalog # 80701-498
Supplier:  ACTON TECHNOLOGIES
Supplier Number:  HGX-10LB
CAS Number:  
Specifications
  • Pack type:
    Plastic drum
  • Description:
    HgX® Mercury Decontaminant Powder
  • Size:
    2 lb.
  • Cat. no.:
    80701-498
  • Supplier No.:
    HGX-10LB
Specifications
About this item
Mercury decontaminant powder reacts rapidly and forming a film over the finely divided, nearly invisible beads of mercury and reacting to produce a non-vaporizing sulfide. It is used for the reduction of toxic mercury vapor.
It is not harmful. None of its components are classified as toxic. It leaves no odor. It has no allergic reactions even after continuous use under various conditions.