Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Marketsource
Supplier: FIREFLYSCI, INC.
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Supplier: FIREFLYSCI, INC.
Marketsource
Certificates
Jump to:
About this item
Multi-Micro Cell Array has 16-chambered multi-micro array of cells for doing multiple measurements.
- Highly accurate results, only need to test sample once
- Made by hand, ensuring high quality optical polishing with no defects
- Fire-fused to ensure that cuvettes are robust and durable
- Comes in UV Quartz
- Available in 10 mm lightpath
- Two windows optically polished
- Walls are made from black quartz to block out stray light
Typically used in Shimadzu machines.
Each cell goes through a rigorous inspection process before leaving the factory.
2 Options Available Below
Product Details & Documents
Multi-Micro Cell Array has 16-chambered multi-micro array of cells for doing multiple measurements.
- Highly accurate results, only need to test sample once
- Made by hand, ensuring high quality optical polishing with no defects
- Fire-fused to ensure that cuvettes are robust and durable
- Comes in UV Quartz
- Available in 10 mm lightpath
- Two windows optically polished
- Walls are made from black quartz to block out stray light
Typically used in Shimadzu machines.
Each cell goes through a rigorous inspection process before leaving the factory.
Product Family Options
Product Information