Orders LinkContactUsLinkComponent
Fixative for hematology, Solution D, (Wescor)
Fixative for hematology, Solution D, (Wescor)
Hematology Stain, Solution D, Fixative (Wescor), 12 × 16 oz.
Catalog # CA10018-834
Supplier:   Azer Scientific
Supplier #:  ES946
Fixative for hematology, Solution D, (Wescor)
Catalog # CA10018-834
Supplier:  Azer Scientific
CAS Number:  
Fixative for hematology, Solution D, (Wescor)
Hematology Stain, Solution D, Fixative (Wescor), 12 × 16 oz.
CAS Number:  
Price
$527.80
Log in for availability
Case of 12 (1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Compatible with the ELITechGroup Aerospray® stainer.
  • Ready to use
  • Lot to lot consistency
A Methanol fixative.
Specifications
Specifications
  • Description:
    Hematology Stain, Solution D, Fixative (Wescor)
  • Size:
    12 × 16 oz.