Sirah Dye Laser Capillary Cells, FireFlySci
Catalog #: 76016-566
Supplier: FIREFLYSCI, INC.
Specifications
- Description:UV Quartz
- Lightpath:6 mm
- Cat. no.:76016-566
Specifications
About this item
Dye laser cells are manufactured with extreme surface flatness extending to the edges of the cell.
- Highly accurate results, only need to test sample once
- Flatness is checked via interferometer
- Made by hand, ensuring high quality optical polishing with no defects
- Fire-fused to ensure that cuvettes are robust and durable
- Available in UV Quartz
- For use in Sirah Dye laser machines
Firefly cuvettes are made with the premium quality quartz and glass material available transmission values.
Each cell goes through a vigorous inspection process before leaving the factory.
Customers Who Bought This Also Bought