Deflagration Spoon
Deflagration Spoon
Catalog # 470313-336
Supplier: GSC International
Supplier #: 4-32901-12
Jump to:
Product Details & Documents
Deflagration Spoons are commonly used in experiments that require students to burn phosphorous, sulfur, or other substances at a safe distance.
The handle is angled to keep hands away from the heat source. It is 18" long with a 3/4" diameter bowl and 1/4" depth.
Specifications
- Description:Deflagration Spoon