Helix® 1070 Series Gloves, HexArmor
Supplier: HexArmor
Certificates
About this item
Helix® 1070 series gloves offer protection yet feel like a second skin, with a smooth finish micro-nitrile coating. Made with a 15-gauge nylon and spandex blended shell, workers will experience excellent comfort and dexterity, allowing hands to move freely and safely on the job.
- High-performance nylon/spandex blend shell offering 360° cut protection
- Microfoam nitrile palm coating provides superior grip and abrasion resistance
- Seamless construction for enhanced comfort and breathability
- Knit wrist helps prevent dirt and debris from entering the glove
- Silicone-free
Helix® series gloves offer a wide variety of grip options to provide users with the appropriate grip for the task at hand. Providing coverage for any working condition, HexArmor is constantly researching and testing new formulas and application methods to maximize grip and function, helping workers redefine safety at their worksite.
Certifications: EN 388 CE 4131X. ANSI/ISEA Cut A1.
Specifications
- Catalog No:
- 76318-046
- 76318-044
- 76318-050
- 76318-048
- 76318-054
- 76318-052
- 76318-040
- 76318-042
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Finish:
- Microfoam Nitrile Palm
- Microfoam Nitrile Palm
- Microfoam Nitrile Palm
- Microfoam Nitrile Palm
- Microfoam Nitrile Palm
- Microfoam Nitrile Palm
- Microfoam Nitrile Palm
- Microfoam Nitrile Palm
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist