Whatman™ Desiccant Pack, Whatman products (Cytiva)
Catalog # CA76312-378
Supplier: Cytiva
Specifications
- Description:Desiccant pack
- Size:1 g
- Cat. no.:CA76312-378
Specifications
About this item
1 g packs of desiccant ensure that DNA sampling cards remain dry during transport or storage.
- High vapor transmission rate
- Substrate is made with Tyvek® material
- Non-dusting, strong and hard to tear