Order Entry
Canada
ContactUsLinkComponent
Capping Reagent 80/20 Acetonitrile/NMI (N-Methylimidazole) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Irritant
Corrosive
Health
Flammable
Capping Reagent 80/20 Acetonitrile/NMI (N-Methylimidazole) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Catalog # CABJBR654-1
Supplier:  Burdick & Jackson
CAS Number:  
Irritant
Corrosive
Health
Flammable
Capping Reagent 80/20 Acetonitrile/NMI (N-Methylimidazole) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Catalog # CABJBR654-1
Supplier:  Burdick & Jackson
Supplier Number:  BR654-1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    2x 1 L
  • Cat. no.:
    CABJBR654-1

Specifications

About this item

 :
Boiling Pt:86.6 °C (main component)
Melting Pt:–43.8 °C (main component)
Density:0.827 g/cm³ (20 °C)
Flash Pt:6 °C (main component)
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS07
 :GHS05
 :GHS08
 :GHS02