Pipette Tip, Tomtec
Catalog # MSPP-196205
Supplier: TOMTEC INC MS
Specifications
- Volume:450 µl
- Cat. no.:MSPP-196205
- Tip Style:Standard orifice
- Supplier No.:196-205
Specifications
About this item
These pipette tips are manufactured with Pro-fax PF511 Polypropylene Homopolymer which is free of slip agents, surfactants, plasticizers, bisphenol A and related agents.