Order Entry
Canada
ContactUsLinkComponent
FURA-2 pentaammonium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
FURA-2 pentaammonium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 89139-200
Supplier:  Biotium
CAS Number:  
FURA-2 pentaammonium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 89139-200
Supplier:  Biotium
Supplier Number:  50030
CAS Number:  

Specifications

  • Size:
    1 mg
  • Cat. no.:
    89139-200
  • Supplier No.:
    50030

Specifications

About this item

 :
 :721 g/mol
Storage Temperature:Refrigerator
Formula:C₂₉H₃₇N₈O₁₄