Fura 2-AM 1 mM in anhydrous DMSO fluorescent indicator, for calcium determination
Catalog # 89139-198
Supplier: Biotium
CAS Number:
Specifications
- Size:1 ml
- Cat. no.:89139-198
- Supplier No.:50029
Specifications
About this item
:
:1001.857 g/mol
Storage Temperature:Freezer
CAS Number:108964-32-5
Formula:C₄₄H₄₇N₃O₂₄
MDL Number:MFCD00036976