Buffer Kit for Calibration of Fluorescent Ca² Indicators, Biotium
Catalog # 89139-296
Supplier: Biotium
CAS Number:
Specifications
- Description:Calcium calibration buffer kit
- Size:2×50 ml (8 assays)
- Cat. no.:89139-296
- Supplier No.:59100
Specifications
About this item
Kit provides a range of calibration buffers with accurate calcium concentrations and is useful for the calibration of fluorescent Ca² indicators (1,2).
- Colorless liquids
- Can be stored at 4 °C
Component A (zero free Ca²): Zero mM CaEGTA (10 mM K₂EGTA, 100 mM KCI and 10 mM MOPS, pH 7,20.
Component B (40 µM free Ca²): 10 mM CaEGTA (10 mM CaEGTA,100 mM KCI and 10 mM MOPS, pH 7,20).
This product contains 50 ml each of components A and B.