Pluronic® F-127
Catalog # 89139-280
Supplier: Biotium
CAS Number:
Specifications
- Size:1 g
- Cat. no.:89139-280
- Supplier No.:59000
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:9003-11-6
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Number:MFCD00082049