About this item
The KOVA-Trol® line of lyophilized, human, urine-based controls provides complete quality control for the physical, chemical and microscopic examination of urine specimens. It is available in three levels to monitor the entire decision range: high abnormal (with or without urobilinogen), low abnormal and normal. Values are assigned for visual and instrument reading on all major systems. It provides the maximum in quality control information for urinalysis testing.
- Red and white cells included for QC of microscopic analysis
- hCG positive in KOVA-Trol® Level III (Normal)
- KOVA-Trol® level II (low abnormal) and KOVA-Trol® III (normal) have microalbumin and creatinine value assigments
- Store at 2 to 8 °C, protect from light
Unreconstituted shelf life is 27 months from date of manufacture. All constituents are stable from day one of reconstitution until day seven with one month frozen stability for strip testing and hCG.