Order Entry
Canada
Orders LinkContactUsLinkComponent
Rotary Scissors, Slice®

Rotary Scissors, Slice®

140 mm, TPE, Stainless Steel 304 , GCr15, White/orange, 16 mm
Catalog # 76314-912
Supplier:   SLICE, INC.
Supplier #:  10598
Rotary Scissors, Slice®
Catalog # 76314-912
Supplier:  SLICE, INC.
Rotary Scissors, Slice®
Rotary Scissors, Slice®

140 mm, TPE, Stainless Steel 304 , GCr15, White/orange, 16 mm

Price
$56.01
Log in for availability
Each Case of 6
Product Details & Documents

The Rotary Scissors by Slice® are an innovative tool to cut through thin, stiff materials quickly and ergonomically.

  • Blade-free scissors
  • Durable reinforced nylon handle with soft-touch grip lining
  • Safer than traditional scissors
  • Ambidextrous design
  • Reduces injuries, lowers costs
  • Oil, lubricant, and maintenance free
  • Lightweight and sturdy

Industrial and home users alike will appreciate the safety of this bladeless tool. Two rotating steel discs feed the material through and shear it cleanly without the user’s fingers ever being in danger. Ideal for paper, gift wrap, retail display cellophane sheets, and other thin, taut materials. Not recommended for flexible materials like plastic wrap or fabric. Designed for right- or left-handed use, the 10598 is comfortable to use, and features a thumb grip and soft-touch liner. Reduce repetitive strain injuries and lower costs with Slice®.

Documents

Specifications
  • Color:
    White/orange
  • Length:
    140 mm
  • Material:
    TPE, Stainless Steel 304 , GCr15
  • Width:
    16 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.