



Hydrogen peroxide 30% stabilized, CMOS for microelectronic use, J.T.Baker®
Catalog # CAJT2201-NM
Supplier: Avantor
CAS Number:
MOC: Regulated
Specifications
- Size:2900 lbs.
- Cat. no.:CAJT2201-NM
Specifications
Specification Test Results
- Assay (H₂O₂):31.52 - 31.72 %
- Free Acid (µeq/g):≤0.2
- Residue after Evaporation:≤10 ppm
- Ammonium (NH₄):≤3 ppm
- Chloride (Cl):≤0.2 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤1 ppm
- Sulfate (SO₄):≤3 ppm
- Trace Impurities - Aluminum (Al):≤47.550 ppb
- Trace Impurities - Antimony (Sb):≤10.000 ppb
- Trace Impurities - Arsenic (As):≤10.000 ppb
- Arsenic and Antimony (as As):≤10.0 ppb
- Trace Impurities - Barium (Ba):≤6.870 ppb
- Trace Impurities - Beryllium (Be):≤10.000 ppb
- Trace Impurities - Bismuth (Bi):≤20.000 ppb
- Trace Impurities - Boron (B):≤6.200 ppb
- Trace Impurities - Cadmium (Cd):≤10.000 ppb
- Trace Impurities - Calcium (Ca):≤10.080 ppb
- Trace Impurities - Chromium (Cr):≤4.930 ppb
- Trace Impurities - Cobalt (Co):≤2.000 ppb
- Trace Impurities - Copper (Cu):≤1.680 ppb
- Trace Impurities - Gallium (Ga):≤20.000 ppb
- Trace Impurities - Germanium (Ge):≤10.000 ppb
- Trace Impurities - Gold (Au):≤10.000 ppb
- Heavy Metals (as Pb):≤500.0 ppb
- Trace Impurities - Iron (Fe):≤13.470 ppb
- Trace Impurities - Lead (Pb):≤2.430 ppb
- Trace Impurities - Lithium (Li):≤1.920 ppb
- Trace Impurities - Magnesium (Mg):≤5.230 ppb
- Trace Impurities - Manganese (Mn):≤1.560 ppb
- Trace Impurities - Molybdenum (Mo):≤10.000 ppb
- Trace Impurities - Nickel (Ni):≤3.260 ppb
- Trace Impurities - Niobium (Nb):≤10.000 ppb
- Trace Impurities - Potassium (K):≤850.000 ppb
- Trace Impurities - Silicon (Si):≤100.000 ppb
- Trace Impurities - Silver (Ag):≤10.000 ppb
- Trace Impurities - Sodium (Na):≤39.700 ppb
- Trace Impurities - Strontium (Sr):≤10.000 ppb
- Trace Impurities - Tantalum (Ta):≤10.000 ppb
- Color (APHA):≤10
- Particle count at point of fill - 0.1 µm and higher (RION KS42A):≤23129.3 par/ml
- Particle count at point of fill - 0.2 µm and higher (RION KS42A):≤1596.8 par/ml
- Particle count at point of fill - 0.5 µm and higher (RION KS42A):≤350.0 par/ml
Specification Test Results
About this item
:
ADR:5.1,II
:34.01
UN:2014
:GHS03
:GHS05
:GHS06
:GHS08
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333