Specifications
- Range:–2600 - 2600 mAbs (Photometric) with 3% FS Accuracy
- Description:MD 100 chlorine, liquid reagents
- Enclosure rating:IP 68
- Wavelength accuracy:±1 nm
- Automatic switch-off:Yes
- Compliance:CE
- Lamp source:LED
- Power supply:4 micro batteries (AAA)
- Weight:260 g
- W×D×H:75×35×155 mm
- Chemical Method:DPD
- Cat. no.:10752-366
- Measurement Range:0.02 - 4.0 mg/l Cl₂
- For Use With:Round Cuvettes 24 mm
- Supplier No.:276005
- Parameters:Chlorine
Specifications
About this item
The MD 100 colorimeter is a single parameter colorimeter for water quality analysis. This line of rugged, waterproof instruments is lightweight and ideal for field testing or for use in the laboratory. This instrument is pre-programmed with three testing ranges for testing Free, Total and Combined Chlorine. All testing ranges use the DPD method and utilize tablet or liquid reagents, depending on the range selected.
- Lightweight, rugged design for use in the field
- Single unit construction with no parts to lose or break
- Large, easy to read display, backlit LCD (on keypress)
- Industry specific parameter combinations available
- The IP 68 design of this instrument ensures that the MD 100 remains water tight for up to one hour at 0.1 m
- The optical system of the utilizes LEDs and interference filters which quickly gives results
- Ergonomic with the rubber feet on the bottom of the housing eliminates the need for two hand operation
A one-point calibration is possible. Additionally have the ability to display readings directly in ABS or %T units.
Automatically stores the last 16 readings with time and date stamp. These results can be transferred to a spreadsheet using the IRiM accessory.
The simple infrared interface allows you to quickly reach the test you need. When the instrument is turned on, it will automatically display the last test used.
Used in chemical, food and beverage, marine, municipalities, NGO, oil and gas, pharmaceutical industries, also in pool public, pool services, power and energy.
Applied in boiler water, cooling water, disinfection control and water treatments.