ContactUsLinkComponent
Versaflo™ Multi-Gas High Efficiency Cartridge, 3M™
Catalog # 76312-270
Supplier:  3M
undefined
Versaflo™ Multi-Gas High Efficiency Cartridge, 3M™
Catalog # 76312-270
Supplier:  3M
Supplier Number:  7100035844
Specifications
  • Color:
    Magenta/Olive
  • Description:
    Multi-Gas High Efficiency Cartridge
  • Cat. no.:
    76312-270
Specifications
About this item
TR-6590N filter is a combination High Efficiency (HE) particulate filter and Multi-Gas cartridge (MG) for use with TR-600 series PAPRs. 3M™ recommends the use of the TR-6500FC filter cover to help protect the filter from physcial damage.
  • High efficiency particulate filter
  • Captures a broad range of certain gases and vapors, including mercury
  • Compatible with the 3M™ Versaflo™ TR-600 PAPR system
  • First multi-gas PAPR cartridge with NIOSH approval for use against mercury
This filter/cartridge is suitable for certain particulates, mercury, certain organic vapors, chlorine dioxide, chlorine, hydrogen chloride, sulfur dioxide, ammonia, methylamine, and/or hydrogen fluoride. This filter/cartridge is NIOSH approved for use against the broadest range of airborne contaminants of any Versaflo™ cartridge.
3M™ recommends the use of the filter cover to help protect the filter/cartridge from physical damage. This filter cover is also used to hold the prefilter or spark arrestor in place.
Certifications: NIOSH approved.