ISO NW Inlet/Exhaust Connector, Welch
80081-226
- Description:Inlet/Exhaust Connector
- Cat. no.:80081-226
- NPT:ISO NW 16
- Thread Size:³/₄-20"
- Supplier No.:1393F
This fitting is made of stainless steel.
- No sealant required for installation
This is specially designed for high vacuum pumps.