Pasteur Pipette, SUN SRi™
Catalog #: 14235-392
Supplier: Thermo Scientific
Specifications
- Capacity:2 ml
- Length:5³/₄"
- Description:Pasteur pipettes
- Cat. no.:14235-392
- Supplier No.:203 770
Specifications
About this item
Pipettes are made of borosilicate glass.
- Straight-cut tips are smooth, fire-polished tops to facilitate bulb attachment
- Without cotton plug
Used in hematology, chemistry, bacteriology as well as general laboratory.
Customers Who Bought This Also Bought