Order Entry
Canada
Orders LinkContactUsLinkComponent
3B Scientific® Absorption Zone

3B Scientific® Absorption Zone

Absorption Zone Of The Root, 1, 43 cm, 43 cm, 8 cm
Catalog # 470311-854
Supplier #:  1002505
3B Scientific® Absorption Zone
Catalog # 470311-854
3B Scientific® Absorption Zone
3B Scientific® Absorption Zone

Absorption Zone Of The Root, 1, 43 cm, 43 cm, 8 cm

Price
$1,132.30
Log in for availability
Each
Product Details & Documents

This relief model shows the absorption zone of a dicotyledonous plant.

The model shows the absorption zone of the root of a white mustard (Sinapis alba) plant.

Specifications
  • Description:
    Absorption Zone Of The Root
  • Height:
    8 cm
  • Length:
    43 cm
  • Width:
    43 cm
  • No. of parts:
    1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.