Measuring Spoon, Hach
About this item
Measuring spoon features a molded plastic cup. Weight determination is based on sodium chloride.
2 Options Available Below
Product Details & Documents
Measuring spoon features a molded plastic cup. Weight determination is based on sodium chloride.
Product Family Options
Product Information
- MaterialCapacityAvailabilityPrice
- Supplier Number: 51100
- Supplier Number: 51000
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)