ProCoat® PVC Dipped Gloves with Interlock Liner, Protective Industrial Products
20000-905
: PIP
- Glove Color:Black
- Ambidextrous/Hand-specific:Hand-specific
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Material:PVC
- Powdered/Powder-free:Powder-free
- Length:14"
- Cuff style:Gauntlet
- Liner material:Cotton
- Finish:Smooth
- Cat. no.:20000-905
PVC coating resists diluted aqueous solutions of bases or acids and provides reliable resistance to oils and organic solvents, making these gloves suitable for mining, construction, refining, petrochemicals and handling oily materials.
- Black color styles hide dirt
- Single dip construction
The interlock lining dries quickly, sheds little lint and offers great dexterity. The straight cuff adds additional length to protect the forearm from liquid run-off.
: EN 388: 4121 mechanical hazards.
: It is strongly advised not to wash and re-use these gloves when they are used to protect against chemical products.