Phone Scope Magnifier, 60×
Catalog # 470310-434
Supplier: L&W OPTICS ELECTRONICS LTD SE
Specifications
- Magnification:60×
- Description:Clip-On Phone Magnifier
- Cat. no.:470310-434
Specifications
About this item
Quick and easy to attach to a phone's camera with the detachable clip.
- 60× Zoom microscope magnifing lens for universal phones
- Bright LED light of the magnifying glass provides enough light at any dim condition
- On/Off switch for LEDs
- Portable and convenient design
- Aligned with GLOBE protocols
This fits many types of cell phones and tablets. Look at images on a cell phone’s large screen, zoom in and take pictures to share with others or input into documents or other media. Quickly attaches to a phone's camera port with the detachable clip.
Perfect for biological, jewelry, coins, currency, stamps, antiques, precision parts, insect and flower photography, geology, horticulture plant identification take up-close pictures of plant issues and send them to others for identification.
Delivery information: LEDs powered by 3 LR1130 cell batteries are included.