Thermal Grip Gloves, Portwest
Supplier: Portwest
Certificates
About this item
Specially designed for use in cold conditions.
- Crinkle Latex coating offers excellent grip
- Palm dipped to increase dexterity and ventilation
- Warm acrylic liner for cold protection
Certifications: AS/NZS 2161 .5;AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 CUT Level (A1). EN 420. EN388:2003 - 1.1.4.2. EN511 - X.1.X.
Specifications
- Catalog No:
- 76103-466
- 76103-468
- 76103-470
- 76103-458
- 76103-460
- 76103-462
- 76103-464
- 76103-456
- Model:
- A140
- A140
- A140
- A140
- A140
- A140
- A140
- A140
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Liner material:
- Acrylic
- Acrylic
- Acrylic
- Acrylic
- Acrylic
- Acrylic
- Acrylic
- Acrylic
- Material:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Finish:
- Palm Dipped
- Palm Dipped
- Palm Dipped
- Palm Dipped
- Palm Dipped
- Palm Dipped
- Palm Dipped
- Palm Dipped
- Gauge:
- 10
- 10
- 10
- 10
- 10
- 10
- 10
- 10