BioClean™ Sterile Neoprene Gloves, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
BioClean Suprene™ sterile neoprene cleanroom gloves are resistant to a wide range of chemicals. Their excellent strength and stretch these gloves are the ideal choice for use within isolators.
- Double-donnable with good grip
- Strong and durable
- Textured fingers and palms
- Non-particulating EasyTear™ packaging
- Powder-free, Latex-free to eliminate Type 1 latex allergies
The hand specific shape and soft neoprene ensure a comfortable and ergonomic fit. Suitable for use in Class 10, ISO 4 or M2.5 and EU GMP Grade A cleanrooms.
Beaded cuff and textured surface for increased tactility and also it comes with excellent ESD properties.
Certifications: In accordance with ASTM D6978. Complies with EN 374-1, 2 & 3:2003 and EN 455-2:2009 glove standards. ISO 9001: 2008 facility. Sterilized in accordance with ISO 11137-1:2006. Conforms to Category 3.Processed in an ISO Class 4 environment. CE 0120 marked.
Specifications
- Catalog No:
- CA75877-712
- CA75877-546
- CA75877-548
- CA75877-552
- CA75877-550
- CA75877-710
- CA75877-708
- Model:
- BSNS
- BSNS
- BSNS
- BSNS
- BSNS
- BSNS
- BSNS
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Neoprene
- Neoprene
- Neoprene
- Neoprene
- Neoprene
- Neoprene
- Neoprene
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Food contact approved:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes