About this item
Estriol ELISA kit is used for the quantification of estradiol in plasma and other sample matrices.
- Measure estradiol (17β-estradiol) in cell culture medium, plasma, and serum
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure estradiol levels down to four pg/ml
- Read: Colorimetric at 405 to 420 nm
Estradiol is the major estrogen secreted by the premenopausal ovary. Estrogens direct the development of the female genotype in embryogenesis and at puberty. In addition, estradiol is an important luteolytic agent in humans.
Plasma estradiol levels increase gradually between days 1 to 7 of the menstrual cycle followed by a sharp increase to a peak value of about 300 pg/ml on day 12 just prior to ovulation. Estradiol levels in most rodent samples are extremely low, rising to detectable levels only during the pro-estrus period and in pregnant females.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Developmental time:90 - 120 min
- Mid point:25 - 60 pg/ml (50% B/B0)
- Detection range:0.9 - 2000 pg/ml
- Storage temperature:–20 °C
- Sensitivity:4 pg/ml (80% B/B0)
- Incubation time:18 hr
Specifications
Related Information
Cayman Chemical Steroid Hormone Assay.pdf
Product Literature
TDS Estriol ELISA Kit