Plug Type Push-in Caps, Simport Scientific
Supplier: Simport Scientific
Certificates
About this item
Plug-type push-in polyethylene caps are designed for test tubes with an outside diameter of 12 mm. The 17 mm caps are designed for T408 centrifuge tubes.
- Anaerobic use or for storage, transfer and centrifuge applications
- Not autoclavable
- Six colors available
Two size options are available for these plug-type caps.
Customers Who Bought This Also Bought