Hajna Tetrathionate (TT) Broth, Hardy Diagnostics
Catalog # 76308-446
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Pack type:Tube
- Description:Hajna TT Broth
- Minimum Order Quantity:5
- Cat. no.:76308-446
- Tube Size:16×125 mm
- Volume:10 ml
Specifications
About this item
For the selective enrichment of Salmonella species from food.
Ordering information: Minimum order must be fiive packs.